Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321173.1 | NP_001308102.1 | 8 | 996 | ||
NM_001321174.1 | NP_001308103.1 | 6 | 832 | |||
NM_001321175.1 | NP_001308104.1 | 9 | 1004 | |||
NM_001321176.1 | NP_001308105.1 | 9 | 999 | |||
NM_005486.2 | NP_005477.2 | 8 | 996 | |||
XM_017024002.1 | XP_016879491.1 | 9 | 916 | |||
GenBank mRNA | AJ010071.1 | - | 8 | 851 | ||
AK001893.1 | - | 8 | 933 | |||
AK223125.1 | - | 8 | 830 | |||
AK300823.1 | - | 6 | 653 | |||
AK303912.1 | - | 7 | 750 | |||
AK303913.1 | - | 8 | 813 | |||
AK309164.1 | - | 6 | 691 | |||
AK315039.1 | - | 8 | 834 | |||
AK315907.1 | - | 6 | 670 | |||
BC029396.1 | - | 8 | 830 | 4612157 |