Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099282.1 | NP_001092752.1 | 4 | 1079 | ||
NM_001099283.1 | NP_001092753.1 | 2 | 916 | |||
NM_001099284.1 | NP_001092754.1 | 3 | 1039 | |||
NM_001324347.1 | NP_001311276.1 | 3 | 957 | |||
NM_001324348.1 | NP_001311277.1 | 2 | 985 | |||
NM_001324349.1 | NP_001311278.1 | 4 | 1149 | |||
NM_001324350.1 | NP_001311279.1 | 3 | 1026 | |||
NM_001324351.1 | NP_001311280.1 | 3 | 1108 | |||
NM_001324352.1 | NP_001311281.1 | 4 | 1084 | |||
NM_001324353.1 | NP_001311282.1 | 5 | 1189 | |||
NM_005674.2 | NP_005665.2 | 2 | 1398 | |||
XM_005271832.2 | XP_005271889.1 | 2 | 911 | |||
XM_006718001.2 | XP_006718064.1 | 2 | 1010 | |||
XM_006718003.3 | XP_006718066.1 | 2 | 910 | |||
XM_011540232.2 | XP_011538534.1 | 3 | 3997 | |||
XM_011540234.2 | XP_011538536.1 | 3 | 1044 | |||
XM_011540235.2 | XP_011538537.1 | 3 | 1043 | |||
XM_011540236.2 | XP_011538538.1 | 3 | 1052 | |||
XM_011540237.2 | XP_011538539.1 | 3 | 1002 | |||
XM_011540238.2 | XP_011538540.1 | 2 | 854 | |||
XM_017016740.1 | XP_016872229.1 | 4 | 1081 | |||
GenBank mRNA | AK292013.1 | - | 3 | 1035 | ||
BC026030.1 | - | 4 | 1023 | 5460088 | ||
X82125.1 | - | 2 | 1398 |