Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003688.1 | NP_001003688.1 | 7 | 1700 | ||
NM_005900.2 | NP_005891.1 | 7 | 1876 | |||
XM_005262992.3 | XP_005263049.1 | 7 | 1669 | |||
XM_006714217.2 | XP_006714280.1 | 7 | 1731 | |||
XM_011531961.1 | XP_011530263.1 | 7 | 1691 | |||
XM_011531962.1 | XP_011530264.1 | 7 | 1736 | |||
XM_011531963.1 | XP_011530265.1 | 7 | 1858 | |||
XM_011531964.1 | XP_011530266.1 | 7 | 1888 | |||
GenBank mRNA | AK096624.1 | - | 3 | 3211 | ||
AK293055.1 | - | 7 | 2002 | |||
BC001878.1 | - | 7 | 1801 | 3546771 | ||
BI772689.1 | - | 328 | ||||
CB956339.1 | - | 1 | 82 | |||
U57456.1 | - | 6 | 1612 | |||
U59423.1 | - | 7 | 1891 | |||
U59912.1 | - | 7 | 1701 |