Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006084.4 | NP_006075.3 | 4, 5 | 615 | ||
GenBank mRNA | AK295467.1 | - | 4, 5 | 508 | ||
AK308347.1 | - | 4, 5 | 538 | |||
BC035716.2 | - | 3, 4 | 491 | 5588434 | ||
M87503.1 | - | 4, 5 | 522 | |||
U92994.1 | - | 2, 3 | 278 |