Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206654.1 | NP_001193583.1 | 46 | 7334 | ||
NM_006312.5 | NP_006303.4 | 47 | 7364 | |||
GenBank mRNA | AF113003.1 | - | 46 | 7053 | ||
AF125672.1 | - | 45 | 7178 | |||
BC004326.1 | - | 8 | 1319 |