Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278270.1 | NP_001265199.1 | 8 | 1287 | ||
NM_006354.3 | NP_006345.1 | 8 | 1893 | |||
NM_133480.2 | NP_597814.1 | 8 | 1893 | |||
NR_103488.1 | 6 | 1329 | ||||
GenBank mRNA | AF069733.1 | - | 7 | 1070 | ||
AK000228.1 | - | 8 | 1297 | |||
AK024982.1 | - | 4 | 1249 | |||
AK292264.1 | - | 8 | 1287 | |||
AL117487.1 | - | 8 | 1556 | |||
BC009240.2 | - | 8 | 1618 | 3009860 | ||
BC013433.1 | - | 8 | 1556 | 3897746 | ||
CR533543.1 | - | 7 | 1070 |