Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006713.3 | NP_006704.3 | 5 | 1218 | ||
XM_011513944.2 | XP_011512246.1 | 6 | 1471 | |||
XM_017008986.1 | XP_016864475.1 | 6 | 1314 | |||
XM_017008987.1 | XP_016864476.1 | 6 | 1191 | |||
GenBank mRNA | AK307268.1 | - | 4 | 471 | ||
BC009610.1 | - | 5 | 1146 | 4297490 | ||
BC010537.1 | - | 5 | 1121 | 3457167 | ||
BC022339.1 | - | 5 | 1151 | 4134117 | ||
BC063402.1 | - | 6 | 1436 | |||
BC071655.1 | - | 5 | 1135 |