Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK314867.1 | - | 2 | 235 | ||
BC020635.1 | - | 2 | 217 | 4717731 | ||
BG482630.1 | - | 2 | 286 | |||
CR749452.1 | - | 2 | 203 | |||
D83920.1 | - | 2 | 191 | |||
DA813504.1 | - | 2 | 329 |