Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007129.3 | NP_009060.2 | 3 | 2903 | ||
GenBank mRNA | AF188733.1 | - | 1 | 1027 | ||
AF193855.1 | - | 3 | 2609 |