Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185063.2 | NP_001171992.1 | 4 | 1748 | ||
NM_007131.4 | NP_009062.2 | 7 | 4353 | |||
XM_005262469.1 | XP_005262526.1 | 7 | 2882 | |||
XM_005262471.3 | XP_005262528.1 | 5 | 3328 | |||
XM_011531391.1 | XP_011529693.1 | 8 | 3069 | |||
XM_011531395.2 | XP_011529697.1 | 4 | 3482 | |||
XM_011531396.2 | XP_011529698.1 | 6 | 3846 | |||
XM_011531397.2 | XP_011529699.1 | 5 | 3754 | |||
XM_017029818.1 | XP_016885307.1 | 6 | 4283 | |||
XM_017029819.1 | XP_016885308.1 | 4 | 7048 | |||
XM_017029820.1 | XP_016885309.1 | 5 | 2261 | |||
GenBank mRNA | AK091687.1 | - | 4 | 1742 | ||
AK092347.1 | - | 4 | 1242 | |||
AK290215.1 | - | 6 | 2057 | |||
AL832801.1 | - | 6 | 1840 | |||
BC109100.2 | - | 6 | 1872 | 40034784 | ||
S67970.1 | - | 2 | 1416 |