Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007360.3 | NP_031386.2 | 8 | 1363 | ||
GenBank mRNA | BC039836.1 | - | 8 | 1327 | 6067176 |
Gene Symbol: KLRC4-KLRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199805.1 | NP_001186734.1 | 13 | 1721 | ||
GenBank mRNA | X54870.1 | - | 10 | 1524 |