Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 2 | 344 | ||
GenBank mRNA | AK225917.1 | - | 2 | 211 | ||
AK315714.1 | - | 2 | 411 | |||
AU122768.1 | - | 2 | 344 | |||
BC020232.1 | - | 2 | 194 | 4652202 | ||
BX248269.1 | - | 2 | 190 | |||
L00635.1 | - | 2 | 198 | |||
L10414.1 | - | 1 | 29 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 3 | 345 | ||
NM_001202559.1 | NP_001189488.1 | 4 | 416 | |||
GenBank mRNA | AK296850.1 | - | 4 | 284 | ||
AK303739.1 | - | 3 | 221 |