Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146108.1 | NP_001139580.1 | 9, 10 | 1150 | ||
NM_012212.3 | NP_036344.2 | 9, 10 | 990 | |||
XM_011518395.2 | XP_011516697.1 | 6, 7 | 826 | |||
GenBank mRNA | AK289597.1 | - | 9, 10 | 1150 | ||
BC035228.2 | - | 9, 10 | 990 | 5122305 | ||
D49387.1 | - | 9, 10 | 920 |