Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018058.2 | NP_001018068.1 | 7 | 1074 | ||
NM_001244583.1 | NP_001231512.1 | 6 | 770 | |||
NM_012252.3 | NP_036384.1 | 8 | 1161 | |||
XM_011515963.1 | XP_011514265.1 | 10 | 1348 | |||
XM_011515964.2 | XP_011514266.1 | 10 | 1363 | |||
XM_011515965.2 | XP_011514267.1 | 10 | 1303 | |||
XM_011515966.2 | XP_011514268.1 | 10 | 1412 | |||
XM_011515969.1 | XP_011514271.1 | 9 | 1291 | |||
XM_011515970.1 | XP_011514272.1 | 9 | 1261 | |||
XM_011515972.2 | XP_011514274.1 | 7 | 877 | |||
XM_017011874.1 | XP_016867363.1 | 9 | 1464 | |||
XM_017011875.1 | XP_016867364.1 | 9 | 1299 | |||
XM_017011876.1 | XP_016867365.1 | 6 | 736 | |||
XM_017011877.1 | XP_016867366.1 | 8 | 1125 | |||
XM_017011878.1 | XP_016867367.1 | 6 | 663 | |||
GenBank mRNA | AK313546.1 | - | 8 | 1113 | ||
BC029891.1 | - | 6 | 1966 | 5179088 | ||
CR933605.1 | - | 7 | 1074 | |||
D43945.1 | - | 8 | 1090 |