Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142556.1 | NP_001136028.1 | 7 | 792 | ||
NM_001142557.1 | NP_001136029.1 | 4 | 565 | |||
NM_012484.2 | NP_036616.2 | 7 | 789 | |||
NM_012485.2 | NP_036617.2 | 6 | 744 | |||
GenBank mRNA | AF032862.1 | - | 7 | 643 | ||
AK290577.1 | - | 6 | 585 | |||
AK302975.1 | - | 8 | 599 | |||
AK303616.1 | - | 4 | 443 | |||
AK313603.1 | - | 7 | 710 | |||
AY623426.1 | - | 7 | 615 | |||
BC017793.1 | - | 6 | 613 | |||
BC030505.2 | - | 7 | 699 | |||
BC057235.1 | - | 6 | 714 | |||
BC070281.1 | - | 6 | 620 | |||
BC108904.1 | - | 7 | 644 | 40007903 | ||
U29343.1 | - | 7 | 670 |