Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002079.2 | NP_002070.1 | 3 | 547 | ||
GenBank mRNA | AF052153.1 | - | 3 | 367 | ||
AK293483.1 | - | 3 | 269 | |||
AK301916.1 | - | 3 | 445 | |||
AK309174.1 | - | 3 | 445 | |||
AK312684.1 | - | 3 | 389 | |||
AY513279.1 | - | 3 | 341 | |||
BC000498.2 | - | 3 | 367 | 2820849 | ||
DA581933.1 | - | 3 | 547 | |||
M37400.1 | - | 3 | 363 |