Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105538.1 | NP_001099008.1 | 17 | 2246 | ||
NM_014520.3 | NP_055335.2 | 17 | 2246 | |||
XM_011523616.2 | XP_011521918.1 | 17 | 2238 | |||
GenBank mRNA | AF147709.1 | - | 17 | 2209 | ||
AL136595.1 | - | 16 | 1954 | |||
BC050546.1 | - | 17 | 2189 | 5495483 |