Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278926.1 | NP_001265855.1 | 9 | 1573 | ||
NM_001278927.1 | NP_001265856.1 | 8 | 1363 | |||
NM_001278929.1 | NP_001265858.1 | 7 | 1433 | |||
NM_015342.3 | NP_056157.1 | 8 | 1539 | |||
XM_005248474.3 | XP_005248531.1 | 4 | 706 | |||
XM_006714583.3 | XP_006714646.1 | 9 | 1509 | |||
XM_011543295.1 | XP_011541597.1 | 7 | 1821 | |||
XM_017009286.1 | XP_016864775.1 | 7 | 863 | |||
GenBank mRNA | AK025679.1 | - | 8 | 1486 | ||
AK298175.1 | - | 8 | 1303 | |||
AK300022.1 | - | 9 | 1516 | |||
AK301654.1 | - | 7 | 1373 | |||
BC015385.1 | - | 8 | 1467 | 4390706 | ||
D38552.1 | - | 8 | 1462 |