Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303241.1 | NP_001290170.1 | 9, 10 | 1186 | ||
XM_005265953.1 | XP_005266010.1 | 8, 9 | 1186 | |||
XM_011534605.2 | XP_011532907.1 | 7, 8 | 1079 | |||
XM_011534606.1 | XP_011532908.1 | 7, 8 | 1094 | |||
XM_011534607.1 | XP_011532909.1 | 8, 9 | 1289 | |||
XM_017009670.1 | XP_016865159.1 | 6, 7 | 971 | |||
XM_017009671.1 | XP_016865160.1 | 6, 7 | 633 | |||
XM_017009672.1 | XP_016865161.1 | 7, 8 | 1094 | |||
GenBank mRNA | AB033020.1 | - | 9, 10 | 1152 | ||
BC152469.1 | - | 9, 10 | 1152 | 8862606 |