Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UIMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199297.2 | NP_001186226.1 | 7 | 1289 | ||
NM_001199298.1 | NP_001186227.1 | 6 | 1064 | |||
NM_016290.4 | NP_057374.3 | 6 | 1078 | |||
XM_005265930.2 | XP_005265987.1 | 6 | 1052 | |||
XM_006714871.2 | XP_006714934.1 | 6 | 1027 | |||
XM_011534568.2 | XP_011532870.1 | 6 | 1043 | |||
XM_011534570.1 | XP_011532872.1 | 6 | 1043 | |||
XM_017009574.1 | XP_016865063.1 | 5 | 943 | |||
GenBank mRNA | AF349313.1 | - | 6 | 1020 | ||
AK023044.1 | - | 7 | 1066 | |||
BC006078.1 | - | 6 | 985 | 3659756 | ||
BX537376.1 | - | 6 | 1078 |