Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001967.3 | NP_001958.2 | 4 | 347 | ||
GenBank mRNA | AB209021.1 | - | 2 | 546 | ||
AK130087.1 | - | 4 | 342 | |||
AK225646.1 | - | 4 | 342 | |||
AY207392.1 | - | 3 | 278 | |||
BC012547.1 | - | 4 | 349 | 4079371 | ||
BC013708.1 | - | 4 | 327 | 4341420 | ||
BC015842.1 | - | 4 | 346 | 4337581 | ||
BC048105.1 | - | 4 | 345 | 4820774 | ||
BT009860.1 | - | 4 | 308 | |||
D30655.1 | - | 4 | 323 | |||
S79942.1 | - | 4 | 323 |