Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SUPT20H SIRNA Assays ›
Gene Symbol:
SUPT20H
Gene Name:
SPT20 homolog, SAGA complex component
Gene Aliases:
C13, C13orf19, FAM48A, FP757, P38IP, SPT20
Chromosome Location:
Chr.13: 37009312 - 37059714 on Build GRCh38
UniGene ID:
Hs.435815
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108969
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SUPT20H
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014286.2
NP_001014308.2
11
1082
NM_001278480.1
NP_001265409.1
11
1082
NM_001278481.1
NP_001265410.1
11
1085
NM_001278482.1
NP_001265411.1
11
1223
NM_017569.3
NP_060039.1
11
1164
XM_005266447.2
XP_005266504.1
11
1058
XM_005266449.3
XP_005266506.1
11
1019
XM_005266451.3
XP_005266508.1
11
1055
XM_005266454.3
XP_005266511.1
10
1017
XM_005266455.2
XP_005266512.1
11
1058
XM_005266456.3
XP_005266513.1
10
1090
XM_005266457.3
XP_005266514.1
10
1016
XM_005266458.3
XP_005266515.1
10
1087
XM_005266460.2
XP_005266517.1
11
1058
XM_005266461.3
XP_005266518.1
11
1058
XM_005266462.4
XP_005266519.1
11
1058
XM_005266464.3
XP_005266521.1
11
1058
XM_005266465.2
XP_005266522.1
11
1055
XM_005266467.3
XP_005266524.1
11
1058
XM_017020653.1
XP_016876142.1
11
1055
XM_017020654.1
XP_016876143.1
10
1017
XM_017020655.1
XP_016876144.1
10
1016
XM_017020656.1
XP_016876145.1
10
1085
XM_017020657.1
XP_016876146.1
11
1055
XM_017020658.1
XP_016876147.1
10
1037
XM_017020659.1
XP_016876148.1
10
1016
XM_017020660.1
XP_016876149.1
11
1055
XM_017020661.1
XP_016876150.1
10
1016
GenBank mRNA
AF093250.1
-
11
1030
AJ130894.1
-
11
1120
AK027554.1
-
10
903
AK225376.1
-
11
1018
AK292376.1
-
11
1168
AK304227.1
-
10
1021
BC001145.1
-
12
1208
3533877
BC030686.2
-
11
1057
4826077
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014170, XM_006232338, XM_006232339, XM_008760995, XM_008760998, and others.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
autophagy
gastrulation
positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0