Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017581.3 | NP_060051.2 | 4 | 854 | ||
GenBank mRNA | AF227732.1 | - | 4 | 697 | ||
AJ243342.1 | - | 4 | 731 | |||
BC113549.1 | - | 4 | 810 | 8322601 | ||
BC113575.1 | - | 4 | 810 | 8322627 |