Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014763.1 | NP_001014763.1 | 2, 3 | 860 | ||
NM_001985.2 | NP_001976.1 | 3, 4 | 475 | |||
GenBank mRNA | AF370381.1 | - | 2, 3 | 842 | ||
AK055285.1 | - | 2, 3 | 862 | |||
AL833205.1 | - | 2, 3 | 3136 | |||
BC093961.1 | - | 3, 4 | 406 | 7939806 | ||
BC093963.1 | - | 3, 4 | 405 | 7939808 | ||
BG281641.1 | - | 2, 3 | 293 | |||
BG399783.1 | - | 3, 4 | 476 | |||
CR456827.1 | - | 3, 4 | 383 | |||
X71129.1 | - | 3, 4 | 410 |