Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195283.1 | NP_001182212.1 | 10 | 2616 | ||
NM_017791.2 | NP_060261.2 | 10 | 3498 | |||
GenBank mRNA | AK000378.1 | - | 10 | 3446 | ||
AY260577.1 | - | 10 | 3478 | |||
CR739521.1 | - | 1 | 444 |