Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136036.2 | NP_001129508.1 | 12 | 2096 | ||
NM_001193328.1 | NP_001180257.1 | 11 | 1648 | |||
NM_017865.3 | NP_060335.2 | 12 | 1880 | |||
XM_011544220.1 | XP_011542522.1 | 12 | 2137 | |||
XM_011544221.1 | XP_011542523.1 | 12 | 2134 | |||
XM_011544222.1 | XP_011542524.1 | 12 | 1752 | |||
XM_011544223.2 | XP_011542525.1 | 11 | 2002 | |||
XM_011544227.1 | XP_011542529.1 | 12 | 2048 | |||
XM_017001698.1 | XP_016857187.1 | 12 | 1749 | |||
XM_017001699.1 | XP_016857188.1 | 11 | 2059 | |||
XM_017001700.1 | XP_016857189.1 | 11 | 1674 | |||
XM_017001701.1 | XP_016857190.1 | 12 | 1864 | |||
XM_017001702.1 | XP_016857191.1 | 11 | 1789 | |||
XM_017001703.1 | XP_016857192.1 | 12 | 2067 | |||
XM_017001704.1 | XP_016857193.1 | 12 | 2024 | |||
XM_017001705.1 | XP_016857194.1 | 12 | 1927 | |||
GenBank mRNA | AB209706.1 | - | 8 | 1934 | ||
AF256175.1 | - | 5 | 1197 | |||
AK000538.1 | - | 11 | 1618 | |||
AK001070.1 | - | 10 | 1297 | |||
AK225535.1 | - | 11 | 1618 | |||
AK302187.1 | - | 12 | 2030 | |||
AL133100.1 | - | 10 | 2609 | |||
BC002948.1 | - | 12 | 1836 | 3533691 | ||
DB520788.1 | - | 1 | 355 |