Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109049
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
5
570
NM_001077352.1
NP_001070820.1
4
522
NM_001308044.1
NP_001294973.1
3
409
NM_018107.4
NP_060577.3
4
522
XM_011536890.2
XP_011535192.1
6
653
XM_011536892.2
XP_011535194.1
5
604
XM_011536893.2
XP_011535195.1
5
605
XM_011536894.2
XP_011535196.1
6
653
XM_011536895.2
XP_011535197.1
5
629
XM_011536896.2
XP_011535198.1
5
605
XM_011536897.2
XP_011535199.1
4
937
XM_011536900.1
XP_011535202.1
5
567
XM_011536902.1
XP_011535204.1
4
419
XM_011536903.1
XP_011535205.1
2
295
XM_011536904.1
XP_011535206.1
4
406
XM_011536905.1
XP_011535207.1
3
404
XM_011536906.1
XP_011535208.1
2
295
XM_017021398.1
XP_016876887.1
5
607
XM_017021399.1
XP_016876888.1
5
608
XM_017021400.1
XP_016876889.1
4
472
XM_017021401.1
XP_016876890.1
5
567
XM_017021402.1
XP_016876891.1
5
478
XM_017021403.1
XP_016876892.1
4
496
XM_017021404.1
XP_016876893.1
4
446
XM_017021405.1
XP_016876894.1
5
567
XM_017021406.1
XP_016876895.1
5
985
XM_017021407.1
XP_016876896.1
5
494
XM_017021408.1
XP_016876897.1
4
484
XM_017021409.1
XP_016876898.1
5
985
XM_017021410.1
XP_016876899.1
4
506
XM_017021411.1
XP_016876900.1
4
937
XM_017021412.1
XP_016876901.1
5
605
XM_017021413.1
XP_016876902.1
4
381
XM_017021414.1
XP_016876903.1
4
419
XM_017021415.1
XP_016876904.1
3
400
XM_017021416.1
XP_016876905.1
2
295
XM_017021417.1
XP_016876906.1
4
406
XM_017021418.1
XP_016876907.1
3
404
XM_017021419.1
XP_016876908.1
2
295
XM_017021420.1
XP_016876909.1
3
824
GenBank mRNA
AF087905.1
-
4
473
AF275678.1
-
5
525
AK001344.1
-
4
472
AK125593.1
-
5
532
AK225792.1
-
5
532
AK295448.1
-
4
484
AK303668.1
-
3
371
AK308055.1
-
4
485
AK315620.1
-
4
484
AL834198.1
-
5
531
BC002566.2
-
4
500
3138909
BC024208.2
-
4
459
2960467
BC106012.1
-
3
323
BX161440.1
-
5
503
CK000110.1
-
4
496
CK003276.1
-
3
401
CR457257.1
-
3
323
DC363343.1
-
4
517
KU178687.1
-
3
323
KU178688.1
-
4
371
KU178689.1
-
3
323
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0