Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093728.2 | NP_001087197.1 | 10 | 1656 | ||
NM_001261817.1 | NP_001248746.1 | 10 | 1641 | |||
NM_001261818.1 | NP_001248747.1 | 9 | 1518 | |||
NM_001261819.1 | NP_001248748.1 | 9 | 1503 | |||
NM_001261820.1 | NP_001248749.1 | 11 | 1862 | |||
NM_001261821.1 | NP_001248750.1 | 11 | 1847 | |||
NM_018679.5 | NP_061149.1 | 10 | 1750 | |||
XM_005249339.2 | XP_005249396.1 | 9 | 1411 | |||
XM_011514830.1 | XP_011513132.1 | 10 | 1907 | |||
XM_011514831.1 | XP_011513133.1 | 8 | 1324 | |||
XM_011514832.1 | XP_011513134.1 | 7 | 1188 | |||
GenBank mRNA | AF260330.1 | - | 7 | 1199 | ||
AF269223.1 | - | 9 | 1434 | |||
AF536532.1 | - | 11 | 1702 | |||
AF536533.1 | - | 10 | 1689 | |||
AK057234.1 | - | 10 | 1473 | |||
AK098590.1 | - | 8 | 1245 | |||
AK301778.1 | - | 10 | 1532 | |||
AK301975.1 | - | 9 | 1439 | |||
AK302022.1 | - | 9 | 1404 | |||
AK302375.1 | - | 10 | 1581 | |||
AK302456.1 | - | 10 | 1562 | |||
AK315078.1 | - | 10 | 1683 | |||
AK316403.1 | - | 10 | 1465 | |||
AY069943.1 | - | 9 | 1458 | |||
BC033729.1 | - | 10 | 1602 | 5167179 | ||
BC040003.1 | - | 11 | 1704 | 5741923 | ||
BC048418.1 | - | 10 | 1680 | 5298037 | ||
BC090050.1 | - | 11 | 1708 | 30387768 |