Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 3 | 397 | ||
GenBank mRNA | AK225917.1 | - | 3 | 264 | ||
AK315714.1 | - | 3 | 464 | |||
AU122768.1 | - | 3 | 397 | |||
BC020232.1 | - | 3 | 247 | 4652202 | ||
BX248269.1 | - | 3 | 243 | |||
L00635.1 | - | 3 | 251 | |||
L10414.1 | - | 2 | 82 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 4 | 398 | ||
NM_001202559.1 | NP_001189488.1 | 5 | 469 | |||
GenBank mRNA | AK296850.1 | - | 5 | 337 | ||
AK303739.1 | - | 4 | 274 |