Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145056.1 | NP_001138528.1 | 22 | 2553 | ||
NM_020428.3 | NP_065161.3 | 22 | 2612 | |||
XM_005259997.1 | XP_005260054.1 | 23 | 2737 | |||
XM_005259999.1 | XP_005260056.1 | 23 | 2678 | |||
GenBank mRNA | AF070636.1 | - | 3 | 613 | ||
AF258549.1 | - | 1 | 99 | |||
AK027519.1 | - | 22 | 2497 | |||
AK075512.1 | - | 11 | 1514 | |||
AK126550.1 | - | 10 | 1302 | |||
AK294118.1 | - | 6 | 906 | |||
AL832517.1 | - | 12 | 2040 | |||
AL832978.1 | - | 19 | 2296 | |||
BC010617.2 | - | 19 | 2303 | |||
BC040556.1 | - | 22 | 2554 | 5268313 | ||
BC042692.1 | - | 22 | 2554 | |||
FN668568.1 | - | 22 | 2612 |