Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127357.1 | NP_001120829.1 | 13 | 1816 | ||
NM_001127358.1 | NP_001120830.1 | 13 | 1893 | |||
NM_020432.4 | NP_065165.3 | 12 | 1804 | |||
XM_005250508.4 | XP_005250565.1 | 14 | 1868 | |||
XM_005250509.4 | XP_005250566.1 | 14 | 1793 | |||
XM_006716063.3 | XP_006716126.1 | 13 | 1739 | |||
XM_011516423.2 | XP_011514725.1 | 13 | 1649 | |||
XM_011516424.2 | XP_011514726.1 | 9 | 2331 | |||
XM_017012459.1 | XP_016867948.1 | 12 | 1674 | |||
GenBank mRNA | AK225854.1 | - | 13 | 1880 | ||
AK308289.1 | - | 13 | 1844 | |||
AL136883.1 | - | 13 | 1784 | |||
AL831969.2 | - | 14 | 1868 | |||
BC127093.1 | - | 13 | 1732 | 40123852 | ||
BC127094.1 | - | 12 | 1720 | 40123853 |