Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022062.2 | NP_071345.2 | 13 | 2660 | ||
XM_005271642.2 | XP_005271699.1 | 12 | 2549 | |||
XM_005271643.2 | XP_005271700.1 | 10 | 2366 | |||
XM_006718894.2 | XP_006718957.1 | 12 | 2436 | |||
XM_011542944.2 | XP_011541246.1 | 13 | 2745 | |||
XM_011542945.2 | XP_011541247.1 | 14 | 2705 | |||
XM_011542946.1 | XP_011541248.1 | 10 | 2280 | |||
XM_011542947.2 | XP_011541249.1 | 11 | 2314 | |||
XM_017018110.1 | XP_016873599.1 | 13 | 5495 | |||
XM_017018111.1 | XP_016873600.1 | 11 | 2355 | |||
XM_017018112.1 | XP_016873601.1 | 7 | 2018 | |||
GenBank mRNA | AB065001.1 | - | 10 | 2339 | ||
AK023136.1 | - | 10 | 2336 | |||
AK023792.1 | - | 3 | 4265 | |||
AL512682.1 | - | 6 | 3487 | |||
BC045626.1 | - | 13 | 2615 | 5261106 | ||
BX648615.1 | - | 11 | 2437 |