Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001755.2 | NP_001746.1 | 6 | 2638 | ||
NM_022845.2 | NP_074036.1 | 6 | 2607 | |||
XM_005256212.3 | XP_005256269.1 | 5 | 2452 | |||
XM_006721321.2 | XP_006721384.1 | 5 | 2452 | |||
XM_017023812.1 | XP_016879301.1 | 5 | 2483 | |||
XM_017023813.1 | XP_016879302.1 | 5 | 2507 | |||
GenBank mRNA | BC018509.1 | - | 6 | 2636 | 3867061 | |
L20298.1 | - | 6 | 2358 |