Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320555.1 | NP_001307484.1 | 4 | 1294 | ||
NM_001320556.1 | NP_001307485.1 | 4 | 1157 | |||
NM_001320557.1 | NP_001307486.1 | 4 | 1302 | |||
NM_025231.2 | NP_079507.1 | 4 | 1244 | |||
XM_017011324.1 | XP_016866813.1 | 4 | 2213 | |||
GenBank mRNA | AK025844.1 | - | 4 | 1191 | ||
BC004255.1 | - | 4 | 1165 | 3609511 |