Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025243.3 | NP_079519.1 | 6 | 2127 | ||
XM_005246874.3 | XP_005246931.1 | 7 | 2290 | |||
XM_011511931.2 | XP_011510233.1 | 6 | 2382 | |||
XM_011511932.1 | XP_011510234.1 | 6 | 2204 | |||
XM_011511933.1 | XP_011510235.1 | 6 | 2470 | |||
XM_017005030.1 | XP_016860519.1 | 7 | 2472 | |||
XM_017005031.1 | XP_016860520.1 | 7 | 2451 | |||
XM_017005032.1 | XP_016860521.1 | 7 | 2332 | |||
XM_017005033.1 | XP_016860522.1 | 7 | 2646 | |||
XM_017005034.1 | XP_016860523.1 | 7 | 2576 | |||
GenBank mRNA | AF283317.1 | - | 6 | 2108 | ||
BC032014.1 | - | 6 | 2082 | 5177540 |