Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109652
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
7
2502
NM_001174093.1
NP_001167564.1
6
2162
NM_001174094.1
NP_001167565.1
7
2499
NM_001174095.1
NP_001167566.1
6
2021
NM_001174096.1
NP_001167567.1
7
2225
NM_001323638.1
NP_001310567.1
11
2872
NM_001323641.1
NP_001310570.1
7
2452
NM_001323642.1
NP_001310571.1
9
2754
NM_001323643.1
NP_001310572.1
9
2532
NM_001323644.1
NP_001310573.1
6
2251
NM_001323645.1
NP_001310574.1
8
2595
NM_001323646.1
NP_001310575.1
10
2833
NM_001323647.1
NP_001310576.1
8
2365
NM_001323648.1
NP_001310577.1
7
2305
NM_001323649.1
NP_001310578.1
8
2368
NM_001323650.1
NP_001310579.1
10
2600
NM_001323651.1
NP_001310580.1
8
2268
NM_001323652.1
NP_001310581.1
9
2417
NM_001323653.1
NP_001310582.1
9
2524
NM_001323654.1
NP_001310583.1
9
2411
NM_001323655.1
NP_001310584.1
9
2521
NM_001323656.1
NP_001310585.1
7
2196
NM_001323657.1
NP_001310586.1
7
2199
NM_001323658.1
NP_001310587.1
7
2038
NM_001323659.1
NP_001310588.1
7
2256
NM_001323660.1
NP_001310589.1
7
2253
NM_001323661.1
NP_001310590.1
8
2399
NM_001323662.1
NP_001310591.1
9
2801
NM_001323663.1
NP_001310592.1
10
2740
NM_001323664.1
NP_001310593.1
9
2724
NM_001323665.1
NP_001310594.1
8
2642
NM_001323666.1
NP_001310595.1
7
2582
NM_001323671.1
NP_001310600.1
8
2478
NM_001323672.1
NP_001310601.1
9
2530
NM_001323673.1
NP_001310602.1
8
2475
NM_001323674.1
NP_001310603.1
5
2000
NM_001323675.1
NP_001310604.1
5
2110
NM_001323676.1
NP_001310605.1
7
2335
NM_001323677.1
NP_001310606.1
7
2332
NM_001323678.1
NP_001310607.1
5
2277
NM_030751.5
NP_110378.3
7
2222
XM_006717498.2
XP_006717561.1
10
2660
XM_011519643.2
XP_011517945.1
10
2963
XM_017016597.1
XP_016872086.1
10
2562
XM_017016598.1
XP_016872087.1
10
5692
XM_017016599.1
XP_016872088.1
9
5530
XM_017016600.1
XP_016872089.1
11
5771
XM_017016601.1
XP_016872090.1
10
5689
XM_017016602.1
XP_016872091.1
11
5768
XM_017016603.1
XP_016872092.1
10
5606
GenBank mRNA
AK091478.1
-
6
2122
AK294775.1
-
7
2417
AK296244.1
-
7
2145
AK299541.1
-
4
1759
AK300830.1
-
6
1969
AK307510.1
-
8
2228
AK314683.1
-
7
2183
AL831979.1
-
9
2480
BC009572.1
-
7
2178
BC112392.1
-
7
2178
40036600
BX647794.1
-
11
2873
M81699.1
-
1
1094
U12170.1
-
7
2183
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0