Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109653
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
9
4043
NM_001174093.1
NP_001167564.1
8
3703
NM_001174094.1
NP_001167565.1
9
4040
NM_001174095.1
NP_001167566.1
8
3562
NM_001174096.1
NP_001167567.1
9
3766
NM_001323638.1
NP_001310567.1
13
4413
NM_001323641.1
NP_001310570.1
9
3993
NM_001323642.1
NP_001310571.1
11
4295
NM_001323643.1
NP_001310572.1
11
4073
NM_001323644.1
NP_001310573.1
8
3792
NM_001323645.1
NP_001310574.1
10
4136
NM_001323646.1
NP_001310575.1
12
4374
NM_001323647.1
NP_001310576.1
10
3906
NM_001323648.1
NP_001310577.1
9
3846
NM_001323649.1
NP_001310578.1
10
3909
NM_001323650.1
NP_001310579.1
12
4141
NM_001323651.1
NP_001310580.1
10
3809
NM_001323652.1
NP_001310581.1
11
3958
NM_001323653.1
NP_001310582.1
11
4065
NM_001323654.1
NP_001310583.1
11
3952
NM_001323655.1
NP_001310584.1
11
4062
NM_001323656.1
NP_001310585.1
9
3737
NM_001323657.1
NP_001310586.1
9
3740
NM_001323658.1
NP_001310587.1
9
3579
NM_001323659.1
NP_001310588.1
9
3797
NM_001323660.1
NP_001310589.1
9
3794
NM_001323661.1
NP_001310590.1
10
3940
NM_001323662.1
NP_001310591.1
11
4342
NM_001323663.1
NP_001310592.1
12
4281
NM_001323664.1
NP_001310593.1
11
4265
NM_001323665.1
NP_001310594.1
10
4183
NM_001323666.1
NP_001310595.1
9
4123
NM_001323671.1
NP_001310600.1
10
4019
NM_001323672.1
NP_001310601.1
11
4071
NM_001323673.1
NP_001310602.1
10
4016
NM_001323674.1
NP_001310603.1
7
3541
NM_001323675.1
NP_001310604.1
7
3651
NM_001323676.1
NP_001310605.1
9
3876
NM_001323677.1
NP_001310606.1
9
3873
NM_001323678.1
NP_001310607.1
7
3818
NM_030751.5
NP_110378.3
9
3763
XM_006717498.2
XP_006717561.1
12
4201
XM_011519643.2
XP_011517945.1
12
4504
XM_017016597.1
XP_016872086.1
12
4103
XM_017016598.1
XP_016872087.1
12
7233
XM_017016599.1
XP_016872088.1
11
7071
XM_017016600.1
XP_016872089.1
13
7312
XM_017016601.1
XP_016872090.1
12
7230
XM_017016602.1
XP_016872091.1
13
7309
XM_017016603.1
XP_016872092.1
12
7147
GenBank mRNA
AK299541.1
-
5
2282
AL831979.1
-
11
4021
BX647794.1
-
13
4416
U12170.1
-
9
3728
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0