Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038618.2 | NP_001033707.1 | 8 | 1102 | ||
NM_001083608.1 | NP_001077077.1 | 7 | 800 | |||
NM_012336.3 | NP_036468.1 | 8 | 944 | |||
NM_031968.2 | NP_114174.1 | 9 | 1082 | |||
XM_005256340.3 | XP_005256397.1 | 8 | 1110 | |||
XM_006722278.3 | XP_006722341.1 | 7 | 897 | |||
XM_011523541.2 | XP_011521843.1 | 8 | 1034 | |||
XM_011523543.2 | XP_011521845.1 | 9 | 947 | |||
XM_017024449.1 | XP_016879938.1 | 9 | 1694 | |||
XM_017024450.1 | XP_016879939.1 | 9 | 1184 | |||
GenBank mRNA | AF128406.1 | - | 8 | 895 | ||
AK000929.1 | - | 8 | 897 | |||
AK056966.1 | - | 8 | 1077 | |||
AK093717.1 | - | 2 | 778 | |||
AK297875.1 | - | 8 | 947 | |||
AK303155.1 | - | 8 | 859 | |||
AL137729.1 | - | 5 | 535 | |||
BC000438.2 | - | 9 | 1049 | 2820621 | ||
BC016440.1 | - | 7 | 687 | 3447710 |