Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032433.2 | NP_115809.1 | 12 | 2348 | ||
XM_005260098.3 | XP_005260155.1 | 13 | 2661 | |||
XM_006722924.2 | XP_006722987.1 | 12 | 2425 | |||
XM_011528362.2 | XP_011526664.1 | 12 | 2389 | |||
XM_011528363.2 | XP_011526665.1 | 14 | 3489 | |||
XM_011528365.2 | XP_011526667.1 | 14 | 2830 | |||
XM_011528366.2 | XP_011526668.1 | 13 | 2658 | |||
XM_011528367.2 | XP_011526669.1 | 13 | 2671 | |||
XM_011528368.2 | XP_011526670.1 | 13 | 2578 | |||
XM_011528369.2 | XP_011526671.1 | 13 | 2591 | |||
XM_011528371.2 | XP_011526673.1 | 6 | 2755 | |||
XM_011528372.2 | XP_011526674.1 | 5 | 1684 | |||
XM_017027367.1 | XP_016882856.1 | 12 | 2365 | |||
XM_017027368.1 | XP_016882857.1 | 11 | 2233 | |||
XM_017027369.1 | XP_016882858.1 | 13 | 3986 | |||
XM_017027370.1 | XP_016882859.1 | 13 | 3999 | |||
XM_017027371.1 | XP_016882860.1 | 13 | 2611 | |||
GenBank mRNA | AB058709.1 | - | 5 | 1662 | ||
AF372702.1 | - | 12 | 2305 | |||
AK094028.1 | - | 13 | 2591 | |||
AK095142.1 | - | 1 | 1230 | |||
AK122817.1 | - | 5 | 2229 | |||
AL832544.1 | - | 6 | 1840 | |||
BX647475.1 | - | 12 | 2309 |