Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033438.3 | NP_254273.2 | 3 | 605 | ||
XM_017002756.1 | XP_016858245.1 | 4 | 2161 | |||
GenBank mRNA | AF275725.1 | - | 3 | 572 | ||
AY034613.1 | - | 3 | 605 | |||
BC074754.2 | - | 3 | 523 | 30915213 |