Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193476.1 | NP_001180405.1 | 16, 17 | 1981 | ||
NM_052961.3 | NP_443193.1 | 16, 17 | 2005 | |||
NM_138718.2 | NP_619732.2 | 14, 15 | 1728 | |||
XM_011514294.2 | XP_011512596.1 | 15, 16 | 2146 | |||
XM_017010235.1 | XP_016865724.1 | 16, 17 | 2031 | |||
GenBank mRNA | AF314959.1 | - | 15, 16 | 1875 | ||
AF331522.1 | - | 16, 17 | 1949 | |||
AK055314.1 | - | 6, 7 | 1451 | |||
AK057276.1 | - | 6, 7 | 524 | |||
AK097410.1 | - | 10, 11 | 1067 | |||
AK122928.1 | - | 16, 17 | 1981 | |||
BC025408.2 | - | 14, 15 | 1721 | 4823098 |