Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080282.3 | NP_525021.3 | 19 | 2933 | ||
GenBank mRNA | AL832004.2 | - | 19 | 2639 | ||
AY028900.1 | - | 19 | 2933 | |||
AY247065.1 | - | 19 | 2964 | |||
BC136460.1 | - | 16 | 2138 | |||
BC144124.1 | - | 16 | 2207 |