Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080284.2 | NP_525023.2 | 27 | 3744 | ||
XM_011524574.2 | XP_011522876.1 | 21 | 3072 | |||
XM_017024404.1 | XP_016879893.1 | 27 | 4056 | |||
XM_017024405.1 | XP_016879894.1 | 27 | 4056 | |||
XM_017024406.1 | XP_016879895.1 | 23 | 3024 | |||
XM_017024407.1 | XP_016879896.1 | 24 | 3128 | |||
XM_017024408.1 | XP_016879897.1 | 13 | 1617 | |||
GenBank mRNA | AF373290.1 | - | 26 | 3572 | ||
AK097296.1 | - | 9 | 1171 | |||
AK125487.1 | - | 6 | 836 | |||
AY028898.1 | - | 27 | 3744 | |||
BC125231.1 | - | 27 | 3743 | |||
BC125232.1 | - | 27 | 3743 |