Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTCFL SIRNA Assays ›
Gene Symbol:
CTCFL
Gene Name:
CCCTC-binding factor like
Gene Aliases:
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
Chromosome Location:
Chr.20: 57495965 - 57525652 on Build GRCh38
UniGene ID:
Hs.131543
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109941
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTCFL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001269040.1
NP_001255969.1
3
882
NM_001269041.1
NP_001255970.1
2
1354
NM_001269042.1
NP_001255971.1
3
882
NM_001269043.1
NP_001255972.1
3
803
NM_001269044.1
NP_001255973.1
2
1354
NM_001269045.1
NP_001255974.1
3
882
NM_001269046.1
NP_001255975.1
3
1328
NM_001269047.1
NP_001255976.1
3
882
NM_001269048.1
NP_001255977.1
2
1354
NM_001269049.1
NP_001255978.1
2
249
NM_001269050.1
NP_001255979.1
2
229
NM_001269051.1
NP_001255980.1
3
882
NM_001269052.1
NP_001255981.1
2
1354
NM_080618.3
NP_542185.2
4
982
NR_072975.1
2
1354
GenBank mRNA
AF336042.1
-
3
781
AK128070.1
-
3
885
AK313990.1
-
3
816
AY071919.1
-
3
809
BC130486.1
-
3
759
40146517
DQ294738.1
-
3
1253
DQ778108.1
-
3
882
DQ778109.1
-
4
982
DQ778110.1
-
2
1354
DQ778111.1
-
3
781
DQ778112.1
-
3
1328
DQ778113.1
-
3
882
DQ778114.1
-
2
1354
DQ778115.2
-
2
1354
DQ778116.2
-
2
1354
DQ778117.2
-
2
1354
DQ778118.2
-
2
1354
DQ778119.2
-
2
1354
DQ778120.2
-
2
1354
DQ778121.3
-
2
1354
DQ778122.2
-
3
882
DQ778123.2
-
3
882
DQ778125.2
-
2
228
DQ778126.2
-
2
227
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of gene expression by genetic imprinting
transcription from RNA polymerase II promoter
cell cycle
positive regulation of gene expression
histone methylation
DNA methylation involved in gamete generation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of histone H3-K4 methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
histone binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline