Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207010.1 | NP_001193939.1 | 7 | 1321 | ||
NM_138337.5 | NP_612210.4 | 6 | 1448 | |||
NM_201623.3 | NP_963917.2 | 5 | 1349 | |||
XM_005253324.2 | XP_005253381.1 | 7 | 1335 | |||
XM_006719033.3 | XP_006719096.1 | 7 | 1316 | |||
XM_011520570.1 | XP_011518872.1 | 8 | 1363 | |||
XM_011520571.2 | XP_011518873.2 | 6 | 1215 | |||
GenBank mRNA | AF247788.1 | - | 6 | 1429 | ||
AY547296.1 | - | 7 | 1321 |