Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRD8 SIRNA Assays ›
Gene Symbol:
BRD8
Gene Name:
bromodomain containing 8
Gene Aliases:
SMAP, SMAP2, p120
Chromosome Location:
Chr.5: 138139766 - 138178669 on Build GRCh38
UniGene ID:
Hs.519337
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
110033
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164326.1
NP_001157798.1
11, 12
1344
NM_001300961.1
NP_001287890.1
11, 12
1340
NM_001300962.1
NP_001287891.1
10, 11
1265
NM_001300966.1
NP_001287895.1
12, 13
1432
NM_006696.3
NP_006687.3
12, 13
1554
NM_139199.1
NP_631938.1
11, 12
1335
XM_005271855.2
XP_005271912.1
12, 13
1545
XM_005271856.2
XP_005271913.1
12, 13
1545
XM_005271857.2
XP_005271914.1
11, 12
1335
XM_005271859.3
XP_005271916.1
10, 11
1116
XM_005271860.3
XP_005271917.1
12, 13
1545
XM_005271861.3
XP_005271918.1
11, 12
1326
XM_011543109.1
XP_011541411.1
12, 13
1545
XM_011543110.1
XP_011541412.1
12, 13
1531
XM_017008969.1
XP_016864458.1
11, 12
1326
XM_017008970.1
XP_016864459.1
12, 13
1545
XM_017008971.1
XP_016864460.1
12, 13
1545
XM_017008972.1
XP_016864461.1
11, 12
1335
XM_017008973.1
XP_016864462.1
11, 12
1326
XM_017008974.1
XP_016864463.1
11, 12
1335
XM_017008975.1
XP_016864464.1
11, 12
1326
XM_017008976.1
XP_016864465.1
10, 11
1116
GenBank mRNA
AB209079.1
-
12, 13
1500
AF016270.1
-
11, 12
1486
AK225928.1
-
11, 12
1307
AK289951.1
-
12, 13
1514
AK293621.1
-
10, 11
1225
AK297608.1
-
2, 3
623
AK297756.1
-
11, 12
1300
AK312333.1
-
12, 13
1514
AL136823.1
-
11, 12
1308
BC008039.2
-
12, 13
1496
3009915
BC008076.2
-
12, 13
1496
3009915
X87613.1
-
11, 12
1362
X94234.1
-
10, 11
1274
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
signal transduction
cell surface receptor signaling pathway
intracellular receptor signaling pathway
regulation of growth
histone H4 acetylation
histone H2A acetylation
transcription factor activity, sequence-specific DNA binding
thyroid hormone receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0