Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152476.2 | NP_689689.2 | 10 | 1061 | ||
XM_011527696.2 | XP_011525998.1 | 10 | 1090 | |||
XM_011527697.2 | XP_011525999.1 | 10 | 1116 | |||
XM_017026327.1 | XP_016881816.1 | 10 | 1070 | |||
XM_017026328.1 | XP_016881817.1 | 10 | 1070 | |||
XM_017026329.1 | XP_016881818.1 | 10 | 1070 | |||
XM_017026330.1 | XP_016881819.1 | 10 | 1017 | |||
GenBank mRNA | AK056548.1 | - | 10 | 1061 | ||
BC101040.1 | - | 10 | 1026 | 40007878 | ||
BC101041.1 | - | 10 | 1026 | 40007880 | ||
BC101042.2 | - | 10 | 1026 | 40007881 |