Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRIM60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258025.1 | NP_001244954.1 | 4 | 1949 | ||
NM_152620.2 | NP_689833.1 | 3 | 1775 | |||
XM_011531683.2 | XP_011529985.1 | 2 | 1769 | |||
GenBank mRNA | AK093201.1 | - | 3 | 1776 |