Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287532.1 | NP_001274461.1 | 3 | 2965 | ||
NM_001287533.1 | NP_001274462.1 | 5 | 3183 | |||
NM_001287534.1 | NP_001274463.1 | 2 | 2838 | |||
NM_007139.3 | NP_009070.2 | 3 | 2934 | |||
NM_152626.3 | NP_689839.1 | 4 | 3061 | |||
GenBank mRNA | AK091618.1 | - | 4 | 2979 | ||
BX647875.1 | - | 2 | 2782 | |||
EU668348.1 | - | 4 | 3005 |