Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152695.5 | NP_689908.3 | 5 | 3483 | ||
XM_011531312.2 | XP_011529614.1 | 6 | 3553 | |||
XM_011531313.2 | XP_011529615.1 | 4 | 4099 | |||
XM_011531314.2 | XP_011529616.1 | 6 | 4240 | |||
XM_017029350.1 | XP_016884839.1 | 5 | 3431 | |||
XM_017029351.1 | XP_016884840.1 | 5 | 3528 | |||
XM_017029352.1 | XP_016884841.1 | 2 | 2731 | |||
GenBank mRNA | AK002078.1 | - | 1 | 1525 | ||
AL832169.1 | - | 1 | 2603 | |||
AL833109.1 | - | 5 | 3442 | |||
AL833407.1 | - | 5 | 3459 | |||
AY280801.1 | - | 5 | 3459 | |||
BC060768.1 | - | 5 | 3460 | 30346713 | ||
BC065938.1 | - | 6 | 3530 | |||
BX537604.1 | - | 2 | 4242 |